Merck
  • The same domain motif for ubiquinone reduction in mitochondrial or chloroplast NADH dehydrogenase and bacterial glucose dehydrogenase.
The same domain motif for ubiquinone reduction in mitochondrial or chloroplast NADH dehydrogenase and bacterial glucose dehydrogenase.
FEBS letters (1990-06-04)
T Friedrich, M Strohdeicher, G Hofhaus, D Preis, H Sahm, H Weiss
ABSTRACT
The respiratory chain NADH:ubiquinone oxidoreductase (NADH dehydrogenase or Complex I) of mitochondria comprises some 30 different subunits, and one FMN and 4 or 5 iron-sulfur clusters as internal redox groups. The bacterial glucose dehydrogenase, which oxidizes glucose to gluconolactone in the periplasmatic space and transfers the electrons to ubiquinone, is a single polypeptide chain with pyrolloquinoline quinone as the only redox group. We report here that the two different enzymes have the same ubiquinone binding domain motif and we discuss the predicted membrane folding of this domain with regard to its role in the proton translocating function of the two enzymes.
Sigma-Aldrich
D-(+)-Gluconic acid δ-lactone, ≥99.0% (GC)
Sigma-Aldrich
Gluconolactone, 99.0-101.0%, meets USP testing specifications