Merck
  • Determination of levoglucosan in atmospheric aerosols using high performance liquid chromatography with aerosol charge detection.
Determination of levoglucosan in atmospheric aerosols using high performance liquid chromatography with aerosol charge detection.
Journal of chromatography. A (2006-02-02)
Roy W Dixon, Gregor Baltzell
ABSTRACT
A sensitive method for analysis of levoglucosan (1,6-anhydro-beta,d-glucopyranose) and other monosaccharide anhydrides, compounds present in biomass combustion smoke, was investigated employing high-performance liquid chromatography (HPLC) with recently developed aerosol charge detection. Aerosol charge detection involves the conversion of the column effluent to an aerosol, which is charged to produce a current. Use of a cation-exchange column and a pure water eluent was found to separate levoglucosan and mannosan from other aerosol components with a detection limit of about 90 ng mL(-1) for levoglucosan or 5 ng injected. This method was demonstrated by successful analysis of aerosol filter samples from three locations.
Sigma-Aldrich
1,6-Anhydro-β-D-glucose, 99%
Supelco
1,6-Anhydro-β-D-glucose, analytical standard