Merck
  • Monitoring of chemical elements and oxidative damage in patients affected by Alzheimer's disease.
Monitoring of chemical elements and oxidative damage in patients affected by Alzheimer's disease.
Annali dell'Istituto superiore di sanita (2005-10-26)
Beatrice Bocca, Giovanni Forte, Francesco Petrucci, Anna Pino, Francesca Marchione, Giuseppe Bomboi, Oreste Senofonte, Franco Giubilei, Alessandro Alimonti
ABSTRACT
The haematic concentration of 26 metals and the oxidative damage in 60 patients (20 males and 40 females) affected by Alzheimer's disease and 44 healthy individuals (33 males and 11 females) were compared. In patients, the following significant (p < or = 0.05) discrepancies were found: i) increment of Ca, Cd, Hg, Mg, Si and Sn, and decrement of Al, Co, Fe and Zn in serum; ii) higher concentrations of Cu, Li, Mn, Sn and Zr and lower of Fe, Hg, Mo in blood; iii) overproduction of oxidant species (SOS) and decrease of the anti-oxidant capacity (SAC) (p < or = 0.001, for both). Variables that, joined, better discriminated between patients and controls resulted to be Si, SOS, SAC, Co, Ca, Al in serum (94% of cases correctly classified) and Cu, Zr, Mo and Fe in blood (90% of cases properly categorized).
Product Description
Supelco
L-Serine, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
L-Glutamic acid, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
L-Serine, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Supelco
L-Phenylalanine, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Sigma-Aldrich
Calcium, dendritic pieces, purified by distillation, 99.9% trace metals basis
Supelco
L-Glutamic acid, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Supelco
L-Isoleucine, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Sigma-Aldrich
Aluminum, foil, thickness 0.13 mm, ≥99.99% trace metals basis
Sigma-Aldrich
Aluminum, pellets, 3-12 mm, 99.99% trace metals basis
Sigma-Aldrich
Aluminum, foil, thickness 0.25 mm, 99.999% trace metals basis
Sigma-Aldrich
Beryllium, chunks, 99.9%
Sigma-Aldrich
Copper, powder (dendritic), <45 μm, 99.7% trace metals basis
Sigma-Aldrich
Copper, foil, thickness 0.25 mm, 99.98% trace metals basis
Sigma-Aldrich
Methylmalonic acid, 99%
Sigma-Aldrich
Titanium, powder, −100 mesh, 99.7% trace metals basis
Sigma-Aldrich
Titanium, sponge, 1-20 mm, 99.5% trace metals basis
Sigma-Aldrich
Zinc, mossy, ≥99%
Sigma-Aldrich
Silicon, pieces, 99.95% trace metals basis
Sigma-Aldrich
Silicon, powder, −325 mesh, 99% trace metals basis
Sigma-Aldrich
Nickel, wire, diam. 0.25 mm, ≥99.9%
Sigma-Aldrich
Bismuth, pieces, 1-12 mm, 99.999% trace metals basis
Supelco
L-Asparagine, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Supelco
L-Isoleucine, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
L-Phenylalanine, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Fumaric acid, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Sigma-Aldrich
Aluminum, foil, thickness 1.0 mm, 99.999% trace metals basis
Sigma-Aldrich
L-Isoleucine, 99%, FCC, FG
Sigma-Aldrich
Titanium, evaporation slug, diam. × L 6.3 mm × 6.3 mm, ≥99.99% trace metals basis
Sigma-Aldrich
Aluminum, foil, thickness 0.45-0.55 mm, 99.999% trace metals basis
Sigma-Aldrich
Copper, powder, <425 μm, 99.5% trace metals basis