Merck
  • Functional characterization of neural and smooth muscle motilin receptors in the chicken proventriculus and ileum.
Functional characterization of neural and smooth muscle motilin receptors in the chicken proventriculus and ileum.
Regulatory peptides (1997-08-15)
T Kitazawa, T Taneike, A Ohga
ABSTRACT
To characterize the motilin receptors present in the chicken, the effects of chicken motilin (Phe-Val-Pro-Phe-Phe-Thr-Gln-Ser-Asp-Ile-Gln-Lys-Met-Gln-Glu-Lys-Glu-Arg -Asn-Lys-Gly-Gln), Leu13 porcine motilin, canine motilin and three erythromycin derivatives (EMA, EM523, GM611) on the contractility of the chicken gastrointestinal (GI) smooth muscles were investigated in vitro and compared with those in the rabbit duodenum. In the proventriculus longitudinal and circular muscle layers, chicken motilin (3 nM-1 microM) caused an atropine- and a tetrodotoxin-sensitive contraction (EC50 = 39-49 nM), and potentiated the EFS-induced contraction without affecting the responsiveness of acetylcholine. EM523 and GM611 (3-100 microM) contracted the proventriculus longitudinal muscle, and the maximum amplitudes of contraction were about 60% of that induced by chicken motilin. Chicken motilin (0.1 nM-100 nM) also caused contraction of the ileum (EC50 = 7 nM) through direct action on the smooth muscle cells. On the other hand, erythromycin derivatives showed only a weak contractile efficacy (about 20% of the maximum response of chicken motilin) even at high concentrations (10-100 microM). The rank order of potency in the ileum was chicken motilin > canine motilin > or = Leu13 porcine motilin > > GM611 > or = EM523 > or = EMA. GM109 slightly inhibited the ideal contractions induced by Leu13 porcine motilin at 100 microM (pA2 = 3.86). In the rabbit duodenum, chicken motilin was a full agonist with the same intrinsic activity as Leu13 porcine motilin, canine motilin and the erythromycin derivatives. However, the rank order of potency (Leu13 porcine motilin > or = canine motilin > chicken motilin > GM611 > or = EM523 > EMA) was different from that in the chicken ileum. In conclusion, chicken motilin causes an excitatory response in the chicken GI tract through activation of neural (proventriculus) and smooth muscle motilin receptors (ileum). The motilin receptor present in the ileum is different from that demonstrated in the rabbit intestine, because of a different rank order of motilin peptides in producing the contraction, low contracting activity of erythromycin derivatives and low antagonistic efficacy of GM109. Different pharmacological characteristics of the mechanical response induced by motilin peptides and erythromycin derivatives between the proventriculus and the ileum are discussed.
Product Description
Supelco
Erythromycin A enol ether, analytical standard