Merck
  • Characterization of thyroglobulin-directed and polyreactive catalytic antibodies in autoimmune disease.
Characterization of thyroglobulin-directed and polyreactive catalytic antibodies in autoimmune disease.
Journal of immunology (Baltimore, Md. : 1950) (1997-08-01)
S Paul, L Li, R Kalaga, J O'Dell, R E Dannenbring, S Swindells, S Hinrichs, P Caturegli, N R Rose
ABSTRACT
Polyreactive and thyroglobulin (Tg)-directed proteolytic activities present in the serum IgG of healthy controls and patients with autoimmune disease were studied by electrophoretic separation of 125I-labeled Tg reaction products and spectrofluorometric measurement of Pro-Phe-Arg-methylcoumarinamide cleavage at the Arg-methylcoumarinamide bond. A decrease of the polyreactive proteolytic activity accompanying an increase of the Tg-cleaving activity in IgG from autoimmune thyroiditis (ATh) and systemic lupus erythematosus (SLE) patients was evident. The Tg, a known target of autoimmune reactions in ATh, was cleaved at lower levels by Abs from patients with this disease than from SLE patients. The Tg-cleaving and Tg-binding activities of the autoantibody preparations were not correlated. Enhanced rates of cleavage at saturating substrate concentrations (Vmax), not increased Tg-binding affinities, were evident in IgG preparations with the greatest Tg-cleaving activity. Similarly, diminution of the polyreactive proteolytic activity in IgG from the autoimmune disease patients was due to decreased Vmax values, not decreased substrate-binding affinities. No cleavage of Tg by IgG from subjects with HIV-1 infection, or from mice hyperimmunized with an albumin-hapten conjugate was evident, suggesting that generation of Tg-cleaving Abs does not accompany V region affinity maturation in response to unrelated Ags. These observations establish Tg as a target of catalytic autoantibodies in SLE and ATh, suggest a transition from polyreactive proteolytic activity to autoantigen-directed activity in autoimmune disease, and open the possibility that combining site chemical reactivity is a factor driving the expression of catalytic activity by autoantibodies.
Sigma-Aldrich
Pro-Phe-Arg-7-amido-4-methylcoumarin, ≥95%
Pricing and availability is not currently available.