Merck
  • Structure-activity relationships of the antimalarial agent artemisinin. 2. Effect of heteroatom substitution at O-11: synthesis and bioassay of N-alkyl-11-aza-9-desmethylartemisinins.
Structure-activity relationships of the antimalarial agent artemisinin. 2. Effect of heteroatom substitution at O-11: synthesis and bioassay of N-alkyl-11-aza-9-desmethylartemisinins.
Journal of medicinal chemistry (1995-12-22)
M A Avery, J D Bonk, W K Chong, S Mehrotra, R Miller, W Milhous, D K Goins, S Venkatesan, C Wyandt, I Khan
ABSTRACT
A novel class of artemisinin analogs, N-alkyl-11-aza-9-desmethylartemisinins 17-29, were synthesized via ozonolysis and acid-catalyzed cyclization of precursor amides 5-16. These amides were prepared through condensation of an activated ester of the known intermediate acid 2 with the corresponding primary amine. The analogs were tested in vitro against W-2 and D-6 strains of Plasmodium falciparum and found in some cases to be more active than artemisinin. A comparison of the in vitro testing methods of Milhous and Makler was conducted and gave similar relative antimalarial activities for these artemisinin analogs. Log P values were determined for most of the compounds, but no apparent correlation between log P and in vitro activity was found.
Sigma-Aldrich
(3R,3aS,6R,6aS,9S,10aS,10bR)-Octahydro-3,6,9-trimethyl-10aH-9,10b-epoxypyrano[4,3,2-jk][2]benzoxepin-2(3H)-one, AldrichCPR