- New enhancers for the chemiluminescent peroxidase catalysed chemiluminescent oxidation of pyrogallol and purpurogallin.
New enhancers for the chemiluminescent peroxidase catalysed chemiluminescent oxidation of pyrogallol and purpurogallin.
The effects of various boronate compounds, 4-biphenylboronic acid, 4-bromobenzene-boronic acid, trans-4-(3-propionic acid)phenylboronic acid and 4-iodophenylboronic acid, on the horseradish peroxidase (HRP) catalysed chemiluminescent oxidation of pyrogallol and purpurogallin by peroxide were investigated. trans-4-(3-Propionic acid)phenylboronic acid produced a 13.7-fold enhancement in the peak light emission from the chemiluminescent HRP catalysed pyrogallol reaction (detection limit for HRP < 1.25 fmol). At low enhancer concentration a single peak of light emission was observed and as the enhancer concentration increased the time to peak light emission became progressively longer. The chemiluminescence showed two peaks at higher concentrations (> 54.3 mumol/L) and the individual peak times depended upon the concentration of the enhancer. All of the boronates enhanced peak light emission in the chemiluminescent HRP catalysed purpurogallin reaction. 4-Biphenylboronic acid was the most effective and it enhanced peak light emission 314-fold. The practical detection limit for HRP (Type VIA) using this enhancer was 4.18 pmol (peak emission at 20 minutes). This compound also enhanced peak light emission 232-fold from a chemiluminescent HRP-purpurogallin reaction in which molecular oxygen replaced peroxide as the oxidant.