Merck
  • Modification by simetryn sulphoxide of a specific thiol group in rat haemoglobin.
Modification by simetryn sulphoxide of a specific thiol group in rat haemoglobin.
The Biochemical journal (1981-10-01)
G J Hughes, C de Jong, R W Fischer, K H Winterhalter, K J Wilson
ABSTRACT
Native rat haemoglobins were found to bind simetryn sulphoxide to an extent 40-fold greater than human haemoglobin. This specific behaviour was studied by using only high-pressure ('performance') liquid chromatography for the preparative separation of globin chains and the isolation of peptides resulting from chemical and enzymic degradation. High recoveries (greater than 80%) of peptides throughout the procedures in combination with microsequence techniques, allow a definitive assignment of the residue undergoing modification. The haemoglobin beta-chain cystine-125 residue, with a stoichiometry of one per tetramer of rat haemoglobin, was found to be modified. Stereochemical implications of this finding are discussed. Simetryn sulphoxide would appear to be useful as a specific reagent for the mapping of exposed thiol residues in proteins.
Product Description
Supelco
Simetryn, PESTANAL®, analytical standard