MilliporeSigma
[Substitutes for oil of cloves in combined histological stains].
Tsitologiia (1986-09-01)
V M Androsova
ABSTRACT
Eugenol basil oil and eugenol are proposed as solvents of dye-stuffs in combined histological staining. These two substances were found to be of full value and more available substitutes of the oil of cloves.