- Continuous-wave InAs/GaAs quantum-dot laser diodes monolithically grown on Si substrate with low threshold current densities.
Continuous-wave InAs/GaAs quantum-dot laser diodes monolithically grown on Si substrate with low threshold current densities.
Optics express (2012-10-06)
Andrew Lee, Qi Jiang, Mingchu Tang, Alwyn Seeds, Huiyun Liu
PMID23037366
ABSTRACT
We report the first room-temperature continuous-wave operation of III-V quantum-dot laser diodes monolithically grown on a Si substrate. Long-wavelength InAs/GaAs quantum-dot structures were fabricated on Ge-on-Si substrates. Room-temperature lasing at a wavelength of 1.28 μm has been achieved with threshold current densities of 163 A/cm(2) and 64.3 A/cm(2) under continuous-wave and pulsed conditions for ridge-waveguide lasers with as cleaved facets, respectively. The value of 64.3 A/cm(2) represents the lowest room-temperature threshold current density for any kind of laser on Si to date.
Sigma-Aldrich
Gallium arsenide, (single crystal substrate), <100>, diam. × thickness 2 in. × 0.5 mm