Merck
Gel mobility shift assays to detect protein-RNA interactions.
Methods in molecular biology (Clifton, N.J.) (2012-06-28)
Alexander V Yakhnin, Helen Yakhnin, Paul Babitzke
ABSTRACT
The gel mobility shift assay is a powerful technique for detecting and quantifying protein-RNA interactions. While other techniques such as filter binding and isothermal titration calorimetry (ITC) are available for quantifying protein-RNA interactions, gel shift analysis provides the added advantage that you can visualize the protein-RNA complexes. In the gel shift assay, protein-RNA complexes are typically separated from the unbound RNA using native polyacrylamide gels in Tris/borate/EDTA buffer, although an alternative Tris-glycine buffering system is superior in many situations. Here, we describe both gel shift methods, along with strategies to improve separation of protein-RNA complexes from free RNA, which can be a particular challenge for small RNA binding proteins.