Merck
Oil migration in chocolate and almond product confectionery systems.
Journal of food science (2012-03-16)
Aylin Altan, David M Lavenson, Michael J McCarthy, Kathryn L McCarthy
ABSTRACT
Oil migration from high oil content almond confections into adjacent chocolate causes changes in product quality. The objective of this study was to quantify the oil migration from almond products to dark chocolate. Magnetic resonance imaging (MRI) was used to monitor spatial and temporal changes of liquid lipid content. A multislice spin echo pulse (MSSE) sequence was used to acquire images with a 7.8-ms echo time and a 1000-ms repetition time using a 1.03T Aspect AI MRI spectrometer. Samples were prepared as a 2-layer model system of chocolate and almond confection. Six different almond products and 1 type of dark chocolate were used. Samples were stored at 20, 25, and 30°C for a time frame of several months. Rate and extent of migration were quantified by a kinetic expression based on the linear dependence of oil uptake by chocolate and the square root of the time. Samples showed distinctly different rate and extent of oil migration, as evidenced by quantitative differences in the kinetic rate constants and equilibrium uptake for the different sample types. This work will be helpful to design formulations for almond and almond-based products in confections. This work will be helpful to design formulations for almond use in confections.