Merck
  • SAR-study on a new class of imidazo[1,2-a]pyridine-based inhibitors of 5-lipoxygenase.
SAR-study on a new class of imidazo[1,2-a]pyridine-based inhibitors of 5-lipoxygenase.
Bioorganic & medicinal chemistry letters (2012-02-14)
Martina Hieke, Carmen B Rödl, Joanna M Wisniewska, Estel la Buscató, Holger Stark, Manfred Schubert-Zsilavecz, Dieter Steinhilber, Bettina Hofmann, Ewgenij Proschak
ABSTRACT
A novel class of 5-lipoxygenase (5-LO) inhibitors characterized by a central imidazo[1,2-a]pyridine scaffold, a cyclohexyl moiety and an aromatic system, is presented. This scaffold was identified in a virtual screening study and exhibits promising inhibitory potential on the 5-LO. Here, we investigate the structure-activity relationships of this compound class. With N-cyclohexyl-6-methyl-2-(4-morpholinophenyl)imidazo[1,2-a]pyridine-3-amine (14), we identified a potent 5-LO inhibitor (IC(50)=0.16μM (intact cells) and 0.1μM (cell-free)), which may possess potential as an effective lead compound intervening with inflammatory diseases and certain types of cancer.
Sigma-Aldrich
Imidazo[1,2-a]pyridine, 99%