Merck
  • Pulmonary arterial hypertension in a patient with Cowden syndrome and anorexigen exposure.
Pulmonary arterial hypertension in a patient with Cowden syndrome and anorexigen exposure.
Chest (2011-10-06)
Delphine Natali, Barbara Girerd, David Montani, Florent Soubrier, Gérald Simonneau, Marc Humbert, Olivier Sitbon
ABSTRACT
We report a case of pulmonary arterial hypertension (PAH) occurring in a patient with Cowden syndrome with a mutation in the phosphatase and tensin (PTEN) tumor suppressor gene, in the context of exposure to the appetite suppressant dexfenfluramine. Anorexigen exposure is known to be a risk factor for PAH. However, the role of PTEN in cell function and the development of pulmonary vascular remodeling and histopathologic signs of PAH in mice with a Pten depletion in smooth muscle cells suggest that the association of PAH and Cowden syndrome may be relevant. In this case report, we hypothesize that PTEN mutations may be a predisposing factor for the development of PAH, with anorexigen exposure as a potential trigger.
Sigma-Aldrich
(+)-Fenfluramine hydrochloride