Merck
  • Allergic contact dermatitis to Myroxylon pereirae (Balsam of Peru) in papaw ointment causing cheilitis.
Allergic contact dermatitis to Myroxylon pereirae (Balsam of Peru) in papaw ointment causing cheilitis.
The Australasian journal of dermatology (2011-08-13)
Stephanie Tan, Mei Mui Tam, Rosemary L Nixon
ABSTRACT
Two patients presented with cheilitis and perioral dermatitis. Patch testing revealed positive reactions to both Myroxylon pereirae (MP; formerly known as Balsam of Peru) and Lucas Papaw Ointment (LPO; Lucas Papaw Remedies, Bisbane, Qld, Australia). MP was not listed as an ingredient in LPO, however the manufacturer confirmed that a small amount of MP was present in LPO and we believe this to be the cause of the patients' dermatitis.