Merck
  • Encapsulation of selenium in chitosan nanoparticles improves selenium availability and protects cells from selenium-induced DNA damage response.
Encapsulation of selenium in chitosan nanoparticles improves selenium availability and protects cells from selenium-induced DNA damage response.
The Journal of nutritional biochemistry (2011-02-05)
Shu Zhang, Yangchao Luo, Huawei Zeng, Qin Wang, Fei Tian, Jiuzhou Song, Wen-Hsing Cheng
ABSTRACT
Selenium, an essential mineral, plays important roles in optimizing human health. Chitosan (CS) is an effective, naturally oriented material for synthesizing nanoparticles with preferable properties such as biocompatibility, biodegradation and resistance to certain enzymes. We have recently shown that cellular exposure to selenium compounds activates ataxia-telangiectasia mutated (ATM)-dependent DNA damage responses, a tumorigenesis barrier. To test whether nanoencapsulation of selenium modulates the cellular response to selenium compounds, the HCT 116 cancerous and the MRC-5 normal cells were treated with Na(2)SeO(3) and methylseleninic acid (MSeA) encapsulated in CS/polyphosphate nanoparticles. Analyses of cellular selenium levels demonstrate that (1) the nanoencapsulation enhances selenium levels in cells after exposure to Na(2)SeO(3) and MSeA (1-10 μM); (2) cells retained more selenium when treated with Na(2)SeO(3) than with MSeA; (3) selenium levels are greater in HCT 116 than in MRC-5 cells after Na(2)SeO(3), but not MSeA, exposure. Survival analysis shows that CS encapsulation desensitizes HCT 116 and MRC-5 cells to Na(2)SeO(3) or MSeA exposure. Immunofluorescent analysis demonstrates that CS encapsulation attenuates the selenium-induced ATM phosphorylation on Ser-1981, and the extent is greater in HCT 116 than in MRC-5 cells. Our results reveal features of selenium nanoencapsulation in CS, including increased selenium retention in cells and decreased cellular sensitivity and DNA damage response to selenium exposure.
Sigma-Aldrich
Methaneseleninic acid, 95%
Pricing and availability is not currently available.