Merck
  • Rapid quantification of tilidine, nortilidine, and bisnortilidine in urine by automated online SPE-LC-MS/MS.
Rapid quantification of tilidine, nortilidine, and bisnortilidine in urine by automated online SPE-LC-MS/MS.
Analytical and bioanalytical chemistry (2010-12-09)
Christoph Köhler, Thomas Grobosch, Torsten Binscheck
ABSTRACT
The opioid tilidine is a prodrug which is hepatically metabolized to active nortilidine and bisnortilidine. Due to the increasing abuse of tilidine by drug users and the lack of a specific immunoassay, we developed an analytical method for the quantification of tilidine, nortilidine, and bisnortilidine in urine suitable for screening. In a following step, this method was used to establish data on excretion kinetics of the substances in order to evaluate the time window of detection after a single oral dose of tilidine/naloxone and also was applied to authentic urine samples from correctional facilities. Urine samples were mixed with internal standard solution and extracted on a weak cation exchanger at pH 6 using a Symbiosis Pico system. The chromatographic separation was achieved within a 3.5-min run time on a Phenylhexyl column (50 × 2.0 mm, 5 μm) via gradient elution (methanol and 0.2% formic acid) at a flow rate of 0.50 mL/min. The ESI-MS/MS was performed on a QTrap 3,200 in positive multiple reaction monitoring mode using two mass transitions per analyte. Validating the method resulted in a lower limit of quantification of 1.0 μg/L followed by a linear calibration range to 100 μg/L for each analyte (r(2) > 0.99). The analytical method allowed the detection of a single dose of a commercially available tilidine solution up to 7 days after administration. Using this highly sensitive method, 55 of 3,665 urine samples were tested positive.
Product Description
Supelco
Tilidine hydrochloride solution, 1.0 mg/mL in methanol (as free base), ampule of 1 mL, certified reference material, Cerilliant®
Pricing and availability is not currently available.