Merck
  • Development of criteria for the detection of adrenosterone administration by gas chromatography-mass spectrometry and gas chromatography-combustion-isotope ratio mass spectrometry for doping control.
Development of criteria for the detection of adrenosterone administration by gas chromatography-mass spectrometry and gas chromatography-combustion-isotope ratio mass spectrometry for doping control.
Drug testing and analysis (2010-04-01)
Lance Brooker, Maria Kristina Parr, Adam Cawley, Ulrich Flenker, Christopher Howe, Rymantas Kazlauskas, Wilhelm Schänzer, Adrian George
ABSTRACT
Adrenosterone (androst-4-ene-3,11,17-trione, 11-oxoandrostenedione) is an endogenous steroid hormone that has been promoted as a dietary supplement capable of reducing body fat and increasing muscle mass. It is proposed that adrenosterone may function as an inhibitor of the 11beta-hydroxysteroid dehydrogenase type 1 enzyme (11beta-HSD1), which is primarily responsible for reactivation of cortisol from cortisone. The urinary metabolism of adrenosterone was investigated, after a single oral administration in two male subjects, by gas chromatography-mass spectrometry (GC-MS) and gas chromatography-combustion-isotope ratio mass spectrometry (GC-C-IRMS). Substantially increased excretion of 11beta-hydroxyandrosterone, 11beta-hydroxyetiocholanolone, 11-oxoandrosterone and 11-oxoetiocholanolone was observed. Minor metabolites such as 3alpha,17beta-dihydroxy-5beta-androstan-11-one, 3alpha-hydroxyandrost-4-ene-11,17-dione and 3alpha,11beta-dihydroxyandrost-4-en-17-one were also identified. The exogenous origin of the most abundant adrenosterone metabolites was confirmed by GC-C-IRMS according to World Anti-Doping Agency criteria. Through analysis of a reference population data set obtained from urine samples provided by elite athlete volunteers (n = 85), GC-MS doping control screening criteria are proposed: 11beta-hydroxyandrosterone concentration greater than 10 000 ng/mL (specific gravity adjusted to 1.020) or 11beta-hydroxyandrosterone/11beta-hydroxyetiocholanolone ratio greater than 20.Urine samples fulfilling these screening criteria may be subjected to GC-C-IRMS analysis for confirmation of adrenosterone administration.
Sigma-Aldrich
Adrenosterone, 98%