Merck
  • The effect of ethanol on oral cocaine pharmacokinetics reveals an unrecognized class of ethanol-mediated drug interactions.
The effect of ethanol on oral cocaine pharmacokinetics reveals an unrecognized class of ethanol-mediated drug interactions.
Drug metabolism and disposition: the biological fate of chemicals (2009-11-19)
Robert B Parker, S Casey Laizure
ABSTRACT
Ethanol decreases the clearance of cocaine by inhibiting the hydrolysis of cocaine to benzoylecgonine and ecgonine methyl ester by carboxylesterases, and there is a large body of literature describing this interaction as it relates to the abuse of cocaine. In this study, we describe the effect of intravenous ethanol on the pharmacokinetics of cocaine after intravenous and oral administration in the dog. The intent is to determine the effect ethanol has on metabolic hydrolysis using cocaine metabolism as a surrogate marker of carboxylesterase activity. Five dogs were administered intravenous cocaine alone, intravenous cocaine after ethanol, oral cocaine alone, and oral cocaine after ethanol on separate study days. Cocaine, benzoylecgonine, and cocaethylene concentrations were determined by high-performance liquid chromatography. Cocaine had poor systemic bioavailability with an area under the plasma concentration-time curve that was approximately 4-fold higher after intravenous than after oral administration. The coadministration of ethanol and cocaine resulted in a 23% decrease in the clearance of intravenous cocaine and a 300% increase in the bioavailability of oral cocaine. Cocaine behaves as a high extraction drug, which undergoes first-pass metabolism in the intestines and liver that is profoundly inhibited by ethanol. We infer from these results that ethanol could inhibit the hydrolysis of other drug compounds subject to hydrolysis by carboxylesterases. Indeed, there are numerous commonly prescribed drugs with significant carboxylesterase-mediated metabolism such as enalapril, lovastatin, irinotecan, clopidogrel, prasugrel, methylphenidate, meperidine, and oseltamivir that may interact with ethanol. The clinical significance of the interaction of ethanol with specific drugs subject to carboxylesterase hydrolysis is not well recognized and has not been adequately studied.
Product Description
Supelco
Cocaethylene solution, 1.0 mg/mL in acetonitrile, ampule of 1 mL, certified reference material, Cerilliant®
Pricing and availability is not currently available.