Merck
  • Regioselective deprotection of orthobenzoates for the synthesis of inositol phosphates.
Regioselective deprotection of orthobenzoates for the synthesis of inositol phosphates.
Organic & biomolecular chemistry (2009-04-04)
Joanna M Swarbrick, Samuel Cooper, Geert Bultynck, Piers R J Gaffney
ABSTRACT
Synthetic myo-inositol 1,4,5-triphosphate, Ins(1,4,5)P(3), and myo-inositol 1,3,4,5-tetraphosphate, Ins(1,3,4,5)P(4), continue to be valuable in biological studies. Inositol orthoesters have proved an important class of intermediate to access these compounds. We investigated the ability of steric bulk from a 4-O protecting group to direct DIBAL-H reduction of inositol orthobenzoates to generate the natural Ins(1,4,5)P(3) precursor 2,3,6-O-tribenzyl myo-inositol. Introduction of an equatorial 4-C-methyl group imparts totally selective reduction and we report the synthesis of novel 4-C-methyl-Ins(1,4,5)P(3) and 4-C-methyl-Ins(1,3,4,5)P(4).
Sigma-Aldrich
D-myo-Inositol 1,3,4,5-tetrakis(phosphate) ammonium salt, ≥95%
Pricing and availability is not currently available.