MilliporeSigma
  • Globotriose-functionalized gold nanoparticles as multivalent probes for Shiga-like toxin.
Globotriose-functionalized gold nanoparticles as multivalent probes for Shiga-like toxin.
Chembiochem : a European journal of chemical biology (2008-04-10)
Yuh-Yih Chien, Mi-Dan Jan, Avijit Kumar Adak, Hsiao-Chien Tzeng, Yen-Ping Lin, Yu-Ju Chen, Ken-Tseng Wang, Chien-Tien Chen, Chia-Chun Chen, Chun-Cheng Lin
ABSTRACT
Compared to monovalent carbohydrates, multivalent carbohydrate ligands exhibit significantly enhanced binding affinities to their interacting proteins. Here, we report globotriose (P(k) ligand)-functionalized gold nanoparticle (AuNP) probes for the investigation of multivalent interactions with the B(5) subunit of Shiga-like toxin I (B-Slt). Six P(k)-ligand-encapsulated AuNPs (P(k)-AuNPs) of varying particle size and linker length were synthesized and evaluated for their potential as multivalent affinity probes by using a surface plasmon resonance competition assay. The affinity of these probes for the interacting proteins was greatly affected by nanoparticle size, linker length, and ligand density on nanoparticle surface. For example, the 20-nm 20-P(k)-l-AuNP, which had a relatively long linker showed a >10(8)-fold increase in affinity compared with the mono P(k) ligand. This intrinsic high-affinity AuNP probe specifically captured the recombinant B-Slt from Escherichia coli lysate, and the resulting purity of the B-Slt was >95 %. We also developed a robust P(k)-AuNP-based detection method for Slt-I by combining the technique with silver enhancement.