Merck
[Studies on triterpenoid constituents in leaf of Ilex pernyi].
Zhongguo Zhong yao za zhi = Zhongguo zhongyao zazhi = China journal of Chinese materia medica (2007-12-07)
Guang-Bo Xie, Si-Xiang Zhou, Lian-Di Lei, Peng-Fei Tu
ABSTRACT
To investigate the chemical constituents of Ilex pernyi. The chemical constituents were isolated by various column chromatographic methods. The structures were identified by spectral data. Eight triterpenoid compounds were isolated and identified as ursolic acid (1), lupeol (2), alpha-amyrin (3), uvaol (4), 3beta-hydroxyurs-11-ene-13beta-olide (5), pomolic acid (6), lup-20 (29)-ene-3beta, 24-diol (7), 3beta, 23-dihydroxy-urs-12-en-28-oic acid (8). The eight compounds were obtained from this plant for the first time.
Sigma-Aldrich
Uvaol, ≥95%
Pricing and availability is not currently available.