Merck
  • A systemic granulomatous response to Schistosoma mansoni eggs alters responsiveness of bone-marrow-derived macrophages to Toll-like receptor agonists.
A systemic granulomatous response to Schistosoma mansoni eggs alters responsiveness of bone-marrow-derived macrophages to Toll-like receptor agonists.
Journal of leukocyte biology (2007-11-22)
Amrita D Joshi, Tracy Raymond, Ana L Coelho, Steven L Kunkel, Cory M Hogaboam
ABSTRACT
Macrophages play a pivotal role in innate and acquired immune responses to Schistosoma mansoni. Classical (M1) or alternative (M2) activation states of these cells further delineate their roles in tissue damage through innate immunity or fibrotic remodeling, respectively. In the present study, we addressed the following question: Does systemic Th2-type cytokine polarization evoked by S. mansoni affect macrophage differentiation and activation? To this end, we analyzed bone marrow-derived macrophages from mice with S. mansoni egg-induced pulmonary granulomas and unchallenged (or naïve) mice to determine their activation state and their response to specific TLR agonists, including S. mansoni egg antigens. Unlike naïve macrophages, macrophages from Th2-polarized mice constitutively expressed significantly higher "found in inflammatory zone-1" (FIZZ1) and ST2 (M2 markers) and significantly lower NO synthase 2, CCL3, MIP-2, TNF-alpha, and IL-12 (M1 markers). Also, compared with naïve macrophages, Th2-polarized macrophages exhibited enhanced responses to the presence of specific TLR agonists, which consistently induced significantly higher levels of gene and protein levels for M2 and M1 markers in these cells. Together, these data show that signals received by bone marrow precursors during S. mansoni egg-induced granuloma responses dynamically alter the development of macrophages and enhance the TLR responsiveness of these cells, which may ultimately have a significant effect on the pulmonary granulomatous response.
Sigma-Aldrich
Pam3-Cys-OH, ≥98.0% (TLC)