Merck
  • Identification of estrogenic/anti-estrogenic compounds in diesel exhaust particulate extract.
Identification of estrogenic/anti-estrogenic compounds in diesel exhaust particulate extract.
Biomedical chromatography : BMC (2007-06-23)
Keiko Noguchi, Akira Toriba, Sang Woon Chung, Ryoichi Kizu, Kazuichi Hayakawa
ABSTRACT
Diesel exhaust particulate extract (DEPE) was obtained from diesel exhaust particulates with Soxhlet extraction using dichloromethane. After separating DEPE into 11 fractions by liquid-liquid extraction, the neutral fraction (N) showed anti-estrogenic activity and the weak acid (phenol) fraction (WA(P)) showed estrogenic and anti-estrogenic activities by a yeast two-hybrid assay system expressing human estrogen receptor alpha. Both fractions were thoroughly fractionated by silica gel column chromatography and reversed-phase HPLC. In the WA(P) fraction, 3-methyl-4-nitrophenol and 2,6-dimethyl-4-nitrophenol were identified by LC-MS/MS as estrogenic compounds. This is the first study to identify 2,6-dimethyl-4-nitrophenol in DEPE and the first study to show that it is an estrogenic compound. In the N fraction, 1-hydroxypyrene was also identified by LC-MS/MS as an anti-estrogenic compound.