Merck
  • Capillary electrophoresis contributions to the hydromorphone metabolism in man.
Capillary electrophoresis contributions to the hydromorphone metabolism in man.
Electrophoresis (2006-05-24)
Andrea Baldacci, Wolfgang Thormann
ABSTRACT
CE-ESI multistage IT-MS (CE-MS(n), n < or = 4) and computer simulation of fragmentation are demonstrated to be effective tools to detect and identify phase I and phase II metabolites of hydromorphone (HMOR) in human urine. Using the same CE conditions as previously developed for the analysis of urinary oxycodone and its metabolites, HMOR and its phase I metabolites produced by N-demethylation, 6-keto-reduction and N-oxidation and phase II conjugates of HMOR and its metabolites formed with glucuronic acid, glucose, and sulfuric acid could be detected in urine samples of a patient that were collected during a pharmacotherapy episode with daily ingestion of 48 mg of HMOR chloride. The CE-MS(n) data obtained with the HMOR standard, synthesized hydromorphol and hydromorphone-N-oxide, and CYP3A4 in vitro produced norhydromorphone were employed to identify the metabolites. This approach led to the identification of previously unknown HMOR metabolites, including HMOR-3O-glucide and various N-oxides, structures for which no standard compounds or mass spectra library data were available. Furthermore, the separation of alpha- and beta-hydromorphol, the stereoisomers of 6-keto-reduced HMOR, was achieved by CE in the presence of the single isomer heptakis(2,3-diacetyl-6-sulfato)-beta-CD. The obtained data indicate that the urinary excretion of alpha-hydromorphol is larger than that of beta-hydromorphol.