Merck
  • LC-ESI-MS/MS analysis for the quantification of morphine, codeine, morphine-3-beta-D-glucuronide, morphine-6-beta-D-glucuronide, and codeine-6-beta-D-glucuronide in human urine.
LC-ESI-MS/MS analysis for the quantification of morphine, codeine, morphine-3-beta-D-glucuronide, morphine-6-beta-D-glucuronide, and codeine-6-beta-D-glucuronide in human urine.
Journal of mass spectrometry : JMS (2005-11-01)
Constance M Murphy, Marilyn A Huestis
ABSTRACT
A liquid chromatographic-electrospray ionization-tandem mass spectrometric method for the quantification of the opiates morphine, codeine, and their metabolites morphine-3-beta-D-glucuronide (M-3-G), morphine-6-beta-D-glucuronide (M-6-G) and codeine-6-beta-D-glucuronide (C-6-G) in human urine has been developed and validated. Identification and quantification were based on the following transitions: 286 to 201 and 229 for morphine, 300 to 215 and 243 for codeine, 462 to 286 [corrected] for M-3-G, 462 to 286 for M-6-G, and 476 to 300 for C-6-G. Calibration by linear regression analysis utilized deuterated internal standards and a weighting factor of 1/X. The method was accurate and precise across a linear dynamic range of 25.0 to 4000.0 ng/ml. Pretreatment of urine specimens using solid phase extraction was sufficient to limit matrix suppression to less than 40% for all five analytes. The method proved to be suitable for the quantification of morphine, codeine, and their metabolites in urine specimens collected from opioid-dependent participants enrolled in a methadone maintenance program.
Product Description
Supelco
Codeine-6-β-D-glucuronide solution, 100 μg/mL in methanol: water (2:8), ampule of 1 mL, certified reference material, Cerilliant®