Merck
Chemical constituents from the leaves of Psidium guajava.
Natural product research (2004-02-27)
Sabira Begum, Syed Imran Hassan, Syed Nawazish Ali, Bina S Siddiqui
ABSTRACT
Five constituents including one new pentacyclic triterpenoid guajanoic acid (1) and four known compounds beta-sitosterol (2), uvaol (3), oleanolic acid (4), and ursolic acid (5) have been isolated from the leaves of Psidium guajava. The new constituent 1 has been characterized as 3beta-p-E-coumaroyloxy-2alpha-methoxyurs-12-en-28-oic acid through 2D NMR techniques and chemical transformations. This is the first report of isolation of compound 3 from the genus Psidium.
Sigma-Aldrich
Uvaol, ≥95%