Merck
  • Enhancement of radiation-induced base release from nucleosides in alkaline solution: essential role of the O.- radical.
Enhancement of radiation-induced base release from nucleosides in alkaline solution: essential role of the O.- radical.
International journal of radiation biology (1992-04-01)
M L Scholes, M N Schuchmann, C von Sonntag
ABSTRACT
The effect of pH on base release in the gamma-radiolysis of N2O-saturated solutions of a number of nucleosides (including uridine, 3-methyluridine, 2',3'-O-isopropylidene-uridine, and adenosine) has been investigated. For all these nucleotides, independent of the base or sugar moiety, base release is very low at pH below 10 (G approximately (0.3-0.7) x 10(-7) mol J-1), but increases drastically to G approximately (3-4) x 10(-7) mol J-1 at pH greater than or equal to 13. This phenomenon had already been previously reported and attributed to an OH(-)-induced transfer of a base radical into a sugar radical. However, it is now shown that at pH 12, where base release starts to increase, a lowering of the dose-rate does not affect the yield of free base. The increase in base release is accompanied by an overall reduction of chromophore loss of similar magnitude (with 2',3'-O-isopropylidene-uridine and 3-methyluridine), as well as by an increase in the yield of oxidizing radicals by a factor of 2 (with uridine). The measured rate constant of the reaction of .OH/O.- with the nucleosides is also pH-dependent, as .OH reacts faster than O.- with the nucleosides by a factor of 6-7. It is concluded that the increase in base release at high pH is caused by the increasing participation of O.-, which, unlike .OH, attacks the nucleosides preferentially at their sugar moieties, and is not due to an OH(-)-induced radical transfer from the base to the sugar moiety.
Sigma-Aldrich
2′,3′-O-Isopropylideneuridine, ≥99% (HPLC)
Pricing and availability is not currently available.