Merck
  • Purification and characterization of beta-methylaspartase from Fusobacterium varium.
Purification and characterization of beta-methylaspartase from Fusobacterium varium.
Molecular and cellular biochemistry (2001-08-17)
S L Bearne, R L White, J E MacDonnell, S Bahrami, J Grønlund
ABSTRACT
Beta-methylaspartase (EC 4.3.1.2) was purified 20-fold in 35% yield from Fusobacterium varium, an obligate anaerobe. The purification steps included heat treatment, fractional precipitation with ammonium sulfate and ethanol, gel filtration, and ion exchange chromatography on DEAE-Sepharose. The enzyme is dimeric, consisting of two identical 46 kDa subunits, and requires Mg2+ (Km = 0.27+/-0.01 mM) and K+ (Km = 3.3+/-0.8 mM) for maximum activity. Beta-methylaspartase-catalyzed addition of ammonia to mesaconate yielded two diastereomeric amino acids, identified by HPLC as (2S,3S)-3-methylaspartate (major product) and (2S,3R)-3-methylaspartate (minor product). Optimal activity for the deamination of (2S,3S)-3-methylaspartate (Km = 0.51+/-0.04 mM) was observed at pH 9.7. The N-terminal protein sequence (30 residues) of the F. varium enzyme is 83% identical to the corresponding sequence of the clostridial enzyme.
Sigma-Aldrich
DL-threo-β-Methylaspartic acid
Pricing and availability is not currently available.