Merck
  • Simultaneous estimation of phenylpropanolamine HCl, guaiphenesin and diphenylpyraline HCl in syrups by LC.
Simultaneous estimation of phenylpropanolamine HCl, guaiphenesin and diphenylpyraline HCl in syrups by LC.
Journal of pharmaceutical and biomedical analysis (2000-12-07)
M Vasudevan, S Ravisankar, A Sathiyanarayanan, R S Chandan
ABSTRACT
A simple, precise and accurate HPLC method was developed for the simultaneous estimation of phenyl-propanolamine HCl, guaiphenesin and diphenylpyraline HCl in syrup. The method was carried out on a Shimpak C8 column with a mobile phase consisting of acetonitrile-triethylamine (pH adjusted to 3.5 using orthophosphoric acid; 0.5%), (35:65, v/v) at a flow rate of 1.2 ml min(-1). Detection was carried out at 210 nm. Diphenhydramine was used as internal standard. The validation of the method was also carried out.
Sigma-Aldrich
4-Diphenylmethoxy-1-methylpiperidine hydrochloride, 95%