Merck
  • Determination of ginsenoside Rf and Rg2 from Panax ginseng using enzyme immunoassay.
Determination of ginsenoside Rf and Rg2 from Panax ginseng using enzyme immunoassay.
Chemical & pharmaceutical bulletin (1998-08-06)
S R Yoon, J J Nah, S K Kim, S C Kim, K Y Nam, D W Jung, S Y Nah
ABSTRACT
We have developed an enzyme immunoassay (EIA) to quantify trace amounts of ginsenoside Rf (Rf), one of the glycosides of protopanaxatriol from Panax ginseng. A carrier protein of bovine serum albumin (BSA) was coupled to the carbohydrate component of Rf using the periodate oxidation method. Antibodies were raised in rabbits using Rf-BSA conjugate as the immunogen and competitive indirect EIA was used for the determination of Rf. The working range was 0.01-10 ng per assay. The anti-Rf antiserum cross-reacted with ginsenoside Rg2 (105%), which is also a component of Panax ginseng and has a very similar chemical structure to Rf. These results suggest that the anti-Rf antiserum could also be used for the quantitation of ginsenoside Rg2 as well as ginsenoside Rf. In a comparison of EIA and HPLC the linear regression equation and correlation coefficient for the two methods were y(EIA) = 1.31x (HPLC)-11.48 and 0.98, respectively.
Product Description
Supelco
Ginsenoside Rf, analytical standard
Ginsenoside Rf, primary reference standard
Supelco
Ginsenoside Rg2, analytical standard