Merck
  • Preparative isolation and purification of harpagoside from Scrophularia ningpoensis hemsley by high-speed counter-current chromatography.
Preparative isolation and purification of harpagoside from Scrophularia ningpoensis hemsley by high-speed counter-current chromatography.
Phytochemical analysis : PCA (2006-12-06)
Shengqiang Tong, Jizhong Yan, Jianzhong Lou
ABSTRACT
The bioactive component harpagoside was successfully separated from the crude extract of Scrophularia ningpoensis Hemsley by one-step purification using high-speed counter-current chromatography (HSCCC). A two-phase solvent system containing n-butanol:ethyl acetate:water (1:9:10) was selected following consideration of the partition coefficient of the target compound. A 276 mg quantity of the crude extract was loaded onto a 250 mL HSCCC column and yielded 11 mg harpagoside at over 97% purity. The chemical structure of harpagoside was determined by HPLC-ESI/MS and 1H-NMR.
Product Description
Supelco
Harpagoside, analytical standard
Pricing and availability is not currently available.
Harpagoside, primary reference standard
Pricing and availability is not currently available.