Merck
  • The pharmacokinetics of clomipramine and desmethylclomipramine in dogs: parameter estimates following a single oral dose and 28 consecutive daily oral doses of clomipramine.
The pharmacokinetics of clomipramine and desmethylclomipramine in dogs: parameter estimates following a single oral dose and 28 consecutive daily oral doses of clomipramine.
Journal of veterinary pharmacology and therapeutics (1998-07-23)
C J Hewson, P D Conlon, U A Luescher, R O Ball
ABSTRACT
Clomipramine is a tricyclic antidepressant that has been recommended for the treatment of canine compulsive disorder. The pharmacokinetics of clomipramine in dogs have not been reported. This study describes the pharmacokinetics of clomipramine and its active metabolite, desmethylclomipramine, in six male dogs. Serial blood samples were collected following both a single oral dose of clomipramine (3 mg/kg) and 28 consecutive daily oral doses (3 mg/kg q 24 h). In addition, 'peak' and 'trough' samples were taken throughout the 28-day dosing period. Plasma was assayed for total (free and protein-bound) clomipramine and desmethylclomipramine, using gas-chromatography with mass spectrometric detection. Various pharmacokinetic parameters were then determined. Following a single dose of clomipramine, time of maximum plasma concentration (tmax) of clomipramine was 0.75-3.1 h, maximum plasma concentration (Cmax) was 16-310 ng/mL and elimination half-life (t1/2el) was 1.2-16 h; tmax of desmethylclomipramine was 1.4-8.8 h, Cmax was 21-134 ng/ mL and t1/2el was 1.2-2.3 h. Following multiple dosing, there was a numeric increase in these parameters; tmax of clomipramine was 3-8 h, Cmax was 43-222 ng/mL and t1/2el was 1.2-16 h; tmax of desmethylclomipramine was 1.4-8.8 h, Cmax was 21-134 ng/mL and t1/2el was 1.2-2.3 h. Clinically significant differences between dogs and humans in the pharmacokinetics of oral clomipramine are discussed.
Product Description
Supelco
N-Desmethylclomipramine hydrochloride solution, 1.0 mg/mL in methanol (as free base), ampule of 1 mL, certified reference material, Cerilliant®
Sigma-Aldrich
Norclomipramine hydrochloride, ≥98% (TLC), powder