Merck
  • Formation of a porphyrin pi-cation radical in the fluoride complex of horseradish peroxidase.
Formation of a porphyrin pi-cation radical in the fluoride complex of horseradish peroxidase.
Biochemistry (1995-11-21)
Z S Farhangrazi, R Sinclair, L Powers, I Yamazaki
ABSTRACT
Horseradish peroxidase (HRP) was oxidized by IrCl6(2-) to a mixture of compounds I and II, the rate of oxidation and the ratio of the mixture being greatly affected by pH (Hayashi & Yamazaki, 1979). Oxidation of HRP by IrCl6(2-) in the presence of fluoride was significantly accelerated. This resulted in the formation of a new compound which is a ferric fluoride complex containing a porphyrin pi-cation radical. The spectrum of the new compound showed a decreased absorption band in the Soret region and a broad band at 570 nm; which was converted to that of the original ferric fluoride complex by addition of ascorbate or hydroquinone. Addition of cyanide slowed down the oxidation of HRP by IrCl6(2-), and the oxidation product was the same as that obtained in the absence of cyanide. Compound I was formed when H2O2 was added to HRP in the presence of fluoride or cyanide. The one-electron reduction potential (Eo') of the oxidized HRP-fluoride complex was measured at several pH values, the Eo' value at pH 7 being 861 +/- 4 mV. The ratio of delta Eo' to delta pH was 49 mV/pH unit.
Sigma-Aldrich
Potassium hexachloroiridate(IV), 99.99% trace metals basis
Sigma-Aldrich
Potassium hexachloroiridate(IV), technical grade