Merck
  • [Durene derivates. IV. N-((2,3,5,6-tetramethylbenzoyl)alkyl)imidazoles and N-((beta-(2,3,5,6-tetramethylphenyl)-beta-hydroxy)ethyl)imidazoles with anticonvulsant activity].
[Durene derivates. IV. N-((2,3,5,6-tetramethylbenzoyl)alkyl)imidazoles and N-((beta-(2,3,5,6-tetramethylphenyl)-beta-hydroxy)ethyl)imidazoles with anticonvulsant activity].
Il Farmaco; edizione scientifica (1981-08-01)
A Leonardi, G Motta, A Subissi, D Nardi
ABSTRACT
A series of N-[(2,3,4,6-tetramethylbenzoyl)alkyl]imidazoles and of N-([beta-(2,3,5,6-tetramethylphenyl)-beta-hydroxy]ethyl)imidazoles has been synthesized. All compounds were tested in mice for anticonvulsant activity against electroshock and pentylenetetrazole convulsions. These durene-derivatives showed good activity, whereas the corresponding benzene-derivatives were inactive.
Product Description
Supelco
1,2,4,5-Tetramethylbenzene, Standard for quantitative NMR, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Sigma-Aldrich
1,2,4,5-Tetramethylbenzene, 98%