Merck
  • Determination of a glucose-containing tetrasaccharide in urine by radioimmunoassay.
Determination of a glucose-containing tetrasaccharide in urine by radioimmunoassay.
Journal of immunological methods (1982-01-01)
D A Zopf, R E Levinson, A Lundblad
ABSTRACT
A radioimmunoassay is described that allows rapid determination of a urinary oligosaccharide -- Glc alpha 1-6Glc alpha 1-4Glc alpha 1-4Glc [(Glc)4] -- at concentrations greater than 2 pmol/microliter. Antibodies produced in rabbits immunized with the phenethylamine derivative of (Glc)4 coupled to keyhole limpet hemocyanin bind tritiated (Glc)4-alditol. Studies comparing the activities of (Glc)4 and several of its derivatives and analogues as inhibitors of binding of tritiated (Glc)4-alditol show that (Glc)4 is detected with maximum specificity and sensitivity only after reduction to (Glc)4-alditol. Quantitation of (Glc)4 in urine samples reduced with sodium borohydride gives excellent agreement with a previously used by more laborious method that employs gas-liquid chromatography-mass spectrometry (GLC/MS) (correlation coefficient = 0.98). Studies of the excretion rate of (Glc)4 in urine of women during pregnancy using the radioimmunoassay method confirm and extend previous results using the GLC/MS method.
Sigma-Aldrich
Glucose tetrasaccharide, ≥99.0% (TLC)
Pricing and availability is not currently available.