Merck
Kinetics and thermodynamics of interfacial transfer.
Journal of pharmaceutical sciences (1983-02-01)
R Fleming, R H Guy, J Hadgraft
ABSTRACT
The kinetic barrier against the transport of methyl and ethyl nicotinates across the water-isopropyl myristate interface has been studied as a function of temperature using a rotating diffusion cell. The temperature dependence of the interfacial transfer kinetics has enabled calculation of the thermodynamic parameters for the process. It is evident from the results that, for the transferring solutes considered, the activation energy barrier is enthalpic, because there is a large positive entropy for transfer into the interfacial region.
Sigma-Aldrich
Ethyl nicotinate, 99%