Merck
Benzofluoranthenes: calculated diol epoxide reactivities.
Chemico-biological interactions (1983-12-01)
B D Silverman, J P Lowe
ABSTRACT
The benzo- and dibenzofluoranthenes, widespread environmental contaminants, have been subjects of continued interest. Calculations of the reactivity of potential ultimate carcinogenic metabolites of the benzofluoranthenes predict unexpected behavior for these non-alternant hydrocarbons.
Product Description
Supelco
Benzo[j]fluoranthene solution, certified reference material, 2000 μg/mL in dichloromethane