Merck
Inhibition of aspartate aminotransferase by hydrazinosuccinate.
Biochimica et biophysica acta (1984-09-07)
R H Yamada, Y Wakabayashi, A Iwashima, T Hasegawa
ABSTRACT
DL-Hydrazinosuccinic acid was synthesized by the reaction of DL-bromosuccinic acid with hydrazine. The compound strongly inhibited aspartate aminotransferase and gave 50% inhibition at 1.3 microM when added simultaneously with L-aspartate to an assay mixture containing enzyme. Incubation of the enzyme with the compound prior to assay resulted in a much stronger inhibition, which proceeded time-dependently. The inhibition was protectable with L-aspartate and was substantially reversed by dialysis.
Sigma-Aldrich
Bromosuccinic acid, 98%