Merck
[Contact allergy to dicyclohexylcarbodiimide and N-hydroxyphthalimide].
Dermatosen in Beruf und Umwelt. Occupation and environment (1987-11-01)
M H Lang, O Hensel
ABSTRACT
We report a case of a female laboratory worker in the chemical industry who acquired contact allergy to dicyclohexylcarbodiimide and N-hydroxyphthalimide. Both substances are used in the chemical synthesis of peptides. So far there have only been a few reports concerning contact allergy to these substances.
Sigma-Aldrich
N-Hydroxyphthalimide, 97%