Merck
  • Diglycolamide-functionalized calix[4]arenes showing unusual complexation of actinide ions in room temperature ionic liquids: role of ligand structure, radiolytic stability, emission spectroscopy, and thermodynamic studies.
Diglycolamide-functionalized calix[4]arenes showing unusual complexation of actinide ions in room temperature ionic liquids: role of ligand structure, radiolytic stability, emission spectroscopy, and thermodynamic studies.
Inorganic chemistry (2013-02-12)
Prasanta K Mohapatra, Arijit Sengupta, Mudassir Iqbal, Jurriaan Huskens, Willem Verboom
ABSTRACT
Diglycolamide-functionalized calix[4]arenes (C4DGAs) with varying structural modifications were evaluated for actinide complexation from their extraction behavior toward actinide ions such as UO2(2+), Pu(4+), PuO2(2+), and Am(3+) in the room temperature ionic liquid (RTIL) 1-n-octyl-3-methylimidazolium bis(trifluoromethane)sulfonamide (C8mimNTf2). The formation constants were calculated for Am(3+) which showed a significant role of ligand structure, nature of substituents, and spacer length. Although the alkyl substituents on the amidic nitrogen increase the extraction efficiency of americium at lower acidity because of the inductive effect of the alkyl groups, at higher acidity the steric crowding around the ligating site determines the extraction efficiency. All C4DGAs formed 1:1 complexes with Am(3+) while for the analogous Eu(3+) complexes no inner sphere water molecules were detected and the asymmetry of the metal ligand complex differed from one another as proved by time-resolved laser induced fluorescence spectroscopy (TRLIFS). Thermodynamic studies indicated that the extraction process, predominant by the Am(3+)-C4DGA complexation reaction, is exothermic. The unique role of the medium on Am(3+) complexation with the C4DGA molecules with varying spacer length, L-IV and L-V, was noticed for the first time with a reversal in the trend observed in the RTIL compared to that seen in a nonpolar molecular diluent like n-dodecane. Various factors leading to a more preorganized structure were responsible for favorable metal ion complexation. The solvent systems show promise to be employed for nuclear waste remediation, and sustainability options were evaluated from radiolytic stability as well as stripping studies.
Sigma-Aldrich
Glycolamide, 98%