Merck
Speed your diagnosis of this gallbladder disorder.
The Journal of family practice (2013-01-18)
David Ivan Croteau
ABSTRACT
Fatty infiltration of the internal organs--and the inflammation associated with it--is an increasingly common cause of gallbladder dysfunction. Here's what to keep in mind to identify it without delay.
Sigma-Aldrich
Iminodiacetic acid, 98%
Sigma-Aldrich
Iminodiacetic acid, purum, ≥98.0% (T)