Merck
  • Thrombin immobilization to methacrylic acid grafted poly(3-hydroxybutyrate) and its in vitro application.
Thrombin immobilization to methacrylic acid grafted poly(3-hydroxybutyrate) and its in vitro application.
Preparative biochemistry & biotechnology (2012-12-12)
Alper Akkaya, Nurdan Pazarlioglu
ABSTRACT
Poly(3-hydroxybutyrate) is nontoxic and biodegradable, with good biocompatibility and potential support for long-term implants. For this reason, it is a good support for enzyme immobilization. Enzyme immobilization could not be done directly because poly(3-hydroxybutyrate) has no functional groups. Therefore, modification should be done for enzyme immobilization. In this study, methacrylic acid was graft polymerized to poly(3-hydroxybutyrate) and thrombin was immobilized to polymethacrylic acid grafted poly(3-hydroxybutyrate). In fact, graft polymerization of methacrylic acid to poly(3-hydroxybutyrate) and thrombin immobilization was a model study. Biomolecule immobilized poly(3-hydroxybutyrate) could be used as an implant. Thrombin was selected as a biomolecule for this model study and it was immobilized to methacrylic acid grafted poly(3-hydroxybutyrate). Then the developed product was used to stop bleeding.
Sigma-Aldrich
Poly(methacrylic acid, sodium salt) solution, average Mw 4,000-6,000, 40 wt. % in H2O
Sigma-Aldrich
Poly[(R)-3-hydroxybutyric acid], natural origin
Sigma-Aldrich
Poly(methacrylic acid, sodium salt) solution, average Mn ~5,400, average Mw ~9,500 by GPC, 30 wt. % in H2O