Merck
  • Clinical trials in spinal cord injury: lessons learned on the path to translation. The 2011 International Spinal Cord Society Sir Ludwig Guttmann Lecture.
Clinical trials in spinal cord injury: lessons learned on the path to translation. The 2011 International Spinal Cord Society Sir Ludwig Guttmann Lecture.
Spinal cord (2012-11-21)
D P Lammertse
ABSTRACT
After three decades of clinical research on interventions to improve neurological outcomes in persons with spinal cord injury (SCI), the promise of preclinical discovery has yet to be translated into a consensus standard of care treatment. Nonetheless, SCI researchers remain hopeful that advances in preclinical discovery coupled with improved clinical trial performance will yield effective restorative treatment. This historical review of key studies in SCI over the past 30 years illustrates the progress that has been achieved in establishing a high standard in the conduct of clinical research while providing important lessons for improving trial design, conduct and reporting. Through application of these lessons, the performance of SCI trials can be improved, thereby shortening the pathway to successful translation and the development of effective therapies.
Sigma-Aldrich
Monosialoganglioside GM1 from bovine brain, lyophilized powder, BioXtra, γ-irradiated, ≥95% (TLC)
Sigma-Aldrich
Monosialoganglioside GM1 from bovine brain, ≥95%, lyophilized powder