Merck
  • Involvement of the sigma1 receptor in the antidepressant-like effects of fluvoxamine in the forced swimming test in comparison with the effects elicited by paroxetine.
Involvement of the sigma1 receptor in the antidepressant-like effects of fluvoxamine in the forced swimming test in comparison with the effects elicited by paroxetine.
European journal of pharmacology (2012-10-09)
Yumi Sugimoto, Noriko Tagawa, Yoshiharu Kobayashi, Kumiko Mitsui-Saito, Yoshihiro Hotta, Jun Yamada
ABSTRACT
We studied the involvement of the sigma(1) receptor in the antidepressant-like effects of the selective serotonin reuptake inhibitor (SSRI) fluvoxamine in DBA/2 mice using the forced swimming test. The effects of the selective sigma(1) receptor antagonist N-[2-(3,4-dichlorophenyl)ethyl]-N-methyl-2-(dimethylamino) ethylamine (BD1047) at 1mg/kg significantly antagonized the anti-immobility elicited by fluvoxamine (10mg/kg). However, the anti-immobility effects elicited by another SSRI, paroxetine (5m/kg), were not altered by BD1047. The selective sigma(1) receptor agonist 2S-(2α,6α,11R(*))-1,2,3,4,5,6-hexahydro-6,11-dimethyl-3-(2-propenyl)-2,6-methano-3-benzazocin-8-ol ((+)SKF-10047) elicited dose-dependent anti-immobility effects in DBA/2 mice. BD1047 significantly blocked the anti-immobility effects induced by (+)SKF-10047 at 10mg/kg. These results suggested that the sigma(1) receptor was associated with fluvoxamine-induced antidepressant-like effects but not with paroxetine-induced antidepressant-like effects.
Product Description
Fluvoxamine for system suitability, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Fluvoxamine maleate, solid