Merck
Calcium with the β-tin structure at high pressure and low temperature.
Proceedings of the National Academy of Sciences of the United States of America (2012-09-27)
Bing Li, Yang Ding, Wenge Yang, Lin Wang, Bo Zou, Jinfu Shu, Stas Sinogeikin, Changyong Park, Guangtian Zou, Ho-kwang Mao
ABSTRACT
Using synchrotron high-pressure X-ray diffraction at cryogenic temperatures, we have established the phase diagram for calcium up to 110 GPa and 5-300 K. We discovered the long-sought for theoretically predicted β-tin structured calcium with I4(1)/amd symmetry at 35 GPa in a s mall low-temperature range below 10 K, thus resolving the enigma of absence of this lowest enthalpy phase. The stability and relations among various distorted simple-cubic phases in the Ca-III region have also been examined and clarified over a wide range of high pressures and low temperatures.
Sigma-Aldrich
Tin, foil, thickness 0.127 mm, 99.9%
Sigma-Aldrich
Tin, powder, 10 μm, 99% trace metals basis
Sigma-Aldrich
Tin, nanopowder, <150 nm particle size (SEM), ≥99% trace metals basis
Sigma-Aldrich
Tin, granular, 0.425-2.0 mm particle size, ≥99.5%, ACS reagent
Sigma-Aldrich
Tin, powder, -100 mesh, 99.99% trace metals basis
Sigma-Aldrich
Tin, powder, <150 μm, 99.5% trace metals basis
Sigma-Aldrich
Tin, ≥99%, powder
Sigma-Aldrich
Tin, foil, thickness 0.5 mm, 99.998% trace metals basis
Sigma-Aldrich
Tin, powder, <45 μm particle size, 99.8% trace metals basis
Sigma-Aldrich
Tin, 99.8%, shot, 3 mm
Sigma-Aldrich
Tin, wire, diam. 0.5 mm, 99.999% trace metals basis
Sigma-Aldrich
Tin, shot, 99.999% trace metals basis