Merck
  • Enantioselective analysis and degradation studies of isocarbophos in soils by chiral liquid chromatography-tandem mass spectrometry.
Enantioselective analysis and degradation studies of isocarbophos in soils by chiral liquid chromatography-tandem mass spectrometry.
Journal of agricultural and food chemistry (2012-09-27)
Hu Zhang, Xinquan Wang, Shulin Zhuang, Nuo Jin, Xiangyun Wang, Mingrong Qian, Hao Xu, Peipei Qi, Qiang Wang, Minghua Wang
ABSTRACT
An enantioselective method is presented for the determination of isocarbophos in soil by liquid chromatography coupled with tandem mass spectrometry. The pesticide residues in soil samples were extracted with acetonitrile, and complete enantioseparation was obtained on an amylose tris(3,5-dimethylphenylcarbamate) chiral column using acetonitrile/2 mM ammonium acetate solution containing 0.1% formic acid (60:40, v/v) as the mobile phase. The absolute configuration of isocarbophos enantiomers was determined by the combination of experimental and calculated electronic circular dichroism spectra. The method was utilized to investigate the degradation of isocarbophos in soils (Changchun, Hangzhou, and Zhengzhou) under sterilized or native conditions. Isocarbophos enantiomers were configurationally stable in the selected soils, and the pesticide degradation was not enantioselective in the sterilized condition. The degradation behavior of rac-isocarbophos was different under native conditions, with no enantioselectivity in the Changchun soil and with the S-(+)-isocarbophos enriched in the Hangzhou and Zhengzhou soils.
Product Description
Supelco
Malathion solution, 100 μg/mL in cyclohexane, PESTANAL®, analytical standard
Supelco
Malathion, PESTANAL®, analytical standard