Merck
  • Effects of fibre-enriched diets on tissue lipid profiles of MSG obese rats.
Effects of fibre-enriched diets on tissue lipid profiles of MSG obese rats.
Food and chemical toxicology : an international journal published for the British Industrial Biological Research Association (2012-08-18)
O A Rotimi, I O Olayiwola, O Ademuyiwa, E A Balogun
ABSTRACT
In order to investigate the influence of some fibre-enriched diets on tissue lipids in an animal model of obesity induced by the administration of monosodium glutamate (MSG), obese rats were fed diets containing 30% of Acha, Cassava, Maize and Plantain for five weeks and weight gain, feed intake and lee index were recorded. The lipid profiles of plasma, erythrocytes, kidney, heart and liver as well as hepatic 3-hydroxyl-3-methylglutaryl-CoA (HMG-CoA) reductase activity were measured. The diets significantly (p<0.05) reduced weight gain and lee index in the obese rats. Obesity-induced increase in plasma and erythrocytes lipid levels was significantly (p<0.05) reduced by these diets. MSG-induced obesity also resulted in a significant increase (p<0.05) in hepatic cholesterol level which was reduced by the diets. MSG-obesity was characterised by a significant (p<0.05) increase in cholesterol, triacylglycerol and phospholipids in kidney and this was reversed by the diets except Maize which did not reverse the increased cholesterol level. Only Acha reversed the obesity-induced increase in heart cholesterol and phospholipids. The increased activity of hepatic HMG-CoA reductase associated with obesity was also significantly (p<0.05) reduced by the diets. In conclusion, dyslipidemia associated with MSG-induced obesity could be attenuated by consumption of fibre-enriched diets.
Sigma-Aldrich
L-Glutamic acid monosodium salt hydrate, ≥99% (HPLC), powder
Sigma-Aldrich
L-Glutamic acid monosodium salt hydrate, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥99%