Merck
  • Ecstacy-induced delayed rhabdomyolysis and neuroleptic malignant syndrome in a patient with a novel variant in the ryanodine receptor type 1 gene.
Ecstacy-induced delayed rhabdomyolysis and neuroleptic malignant syndrome in a patient with a novel variant in the ryanodine receptor type 1 gene.
Anaesthesia (2012-06-28)
T Russell, S Riazi, N Kraeva, A C Steel, L A Hawryluck
ABSTRACT
We present the case of a 20-year-old woman who developed rhabdomyolysis, disseminated intravascular coagulopathy and multi-organ failure induced by ecstasy. Following initial improvement, she developed delayed rhabdomyolysis then haloperidol-induced neuroleptic malignant syndrome, which was treated with a total of 50 mg.kg(-1) dantrolene. Subsequent genetic testing revealed a novel potentially pathogenic variant in the ryanodine receptor type 1 gene. However, caffeine-halothane contracture testing of the patient's mother who carried the same gene variant was negative for malignant hyperthermia.
Sigma-Aldrich
2-Bromo-2-chloro-1,1,1-trifluoroethane, ≥99%
Pricing and availability is not currently available.