Merck
  • Novel molecular mechanisms of antitumor action of dichloroacetate against T cell lymphoma: Implication of altered glucose metabolism, pH homeostasis and cell survival regulation.
Novel molecular mechanisms of antitumor action of dichloroacetate against T cell lymphoma: Implication of altered glucose metabolism, pH homeostasis and cell survival regulation.
Chemico-biological interactions (2012-06-19)
Ajay Kumar, Shiva Kant, Sukh Mahendra Singh
ABSTRACT
Pyruvate dehydrogenase kinase (PDK) inhibits pyruvate dehydrogenase (PDH) activity and thus promotes energetic switch from mitochondrial glucose oxidation to cytoplasmic glycolysis in cancerous cells (a phenomenon known as the 'Warburg effect') for their energy need, which facilitates the cancer progression by resisting induction of apoptosis and promoting tumor metastasis. Thus, in the present investigation, we explored the molecular mechanisms of the tumoricidal action of dichloroacetate (DCA), a pyruvate dehydrogenase kinase inhibitor, on cells of a murine T cell lymphoma, designated as Dalton's lymphoma (DL). In vitro treatment of tumor cells with DCA inhibited their survival accompanied by a modulation of the biophysical composition of tumor-conditioned medium with respect to pH, glucose and lactate. DCA treatment also altered expression of HIF1-α and pH regulators: VATPase and MCT1 and production of cytokines: IL-10, IL-6 and IFN-γ. Moreover, we also observed an alteration in the expression of other apoptosis and cell survival regulatory molecules: PUMA, GLUT1, Bcl2, p53, CAD, caspase-3 and HSP70. The study discusses the role of novel molecular mechanisms underlying DCA-dependent inhibition of tumor cell survival. This study shows for the first time that DCA-dependent alteration of tumor cell survival involves altered pH homeostasis and glucose metabolism. Thus, these findings will provide a new insight for therapeutic applications of DCA as a novel antineoplastic agent against T cell lymphoma.
Sigma-Aldrich
Dichloroacetic acid, ReagentPlus®, ≥99%
Supelco
Dichloroacetic acid, PESTANAL®, analytical standard
Sigma-Aldrich
DCA Deblock (0.36M dichloroacetic acid in toluene)